This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00521603_m1
See other FAM129B GE Assays ›
Gene Symbol:
Fam129b
Gene Name:
family with sequence similarity 129, member B
Gene Aliases:
9130404D14Rik
Chromosome Location:
Chr.2: 32876134 - 32925253 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00521603_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fam129b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146119.2 NP_666231.1 4 - 5 635 76
GenBank mRNA AK046583.1 - 4 - 5 532 76
AK051396.1 - 4 - 5 574 76
AK146953.1 - 4 - 5 620 76
AK149818.1 - 4 - 5 546 76
AK149888.1 - 4 - 5 545 76
AK150642.1 - 4 - 5 573 76
AK152207.1 - 4 - 5 594 76
AK153489.1 - 4 - 5 579 76
AK170512.1 - 4 - 5 575 76
BC027843.1 - 4 - 5 555 5343013 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200000597.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fam129b

Gene Ontology Categories:

Function(s)

negative regulation of apoptotic process

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200005498.mm.1 NM_146119.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200005498.mm.1 NM_146119.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200005498.mm.1 TC0200000597.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0