This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00518349_m1
See other DHRS1 GE Assays ›
Gene Symbol:
Dhrs1
Gene Name:
dehydrogenase/reductase (SDR family) member 1
Gene Aliases:
1110029G07Rik, AW112170, D14Ertd484e
Chromosome Location:
Chr.14: 55739020 - 55745684 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00518349_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dhrs1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026819.3 NP_081095.2 2 - 3 417 73
GenBank mRNA AK003958.1 - 2 - 3 307 73
AK146317.1 - 2 - 3 364 73
AK159964.1 - 2 - 3 339 73
AK168115.1 - 2 - 3 364 73
AK168147.1 - 2 - 3 364 73
BC003930.1 - 2 - 3 348 3485584 73
BU963142.1 - 420 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002100.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dhrs1

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
metabolic process
oxidation-reduction process
molecular_function
oxidoreductase activity
pinocarveol dehydrogenase activity
chloral hydrate dehydrogenase activity
hydroxymethylmethylsilanediol oxidase activity
1-phenylethanol dehydrogenase activity
myrtenol dehydrogenase activity
cis-1,2-dihydroxy-1,2-dihydro-8-carboxynaphthalene dehydrogenase activity
3-hydroxy-4-methyloctanoyl-CoA dehydrogenase activity
2-hydroxy-4-isopropenylcyclohexane-1-carboxyl-CoA dehydrogenase activity
cis-9,10-dihydroanthracene-9,10-diol dehydrogenase activity
citronellol dehydrogenase activity
naphthyl-2-hydroxymethyl-succinyl-CoA dehydrogenase activity
2,4,4-trimethyl-1-pentanol dehydrogenase activity
2,4,4-trimethyl-3-hydroxypentanoyl-CoA dehydrogenase activity
1-hydroxy-4,4-dimethylpentan-3-one dehydrogenase activity
endosulfan diol dehydrogenase activity
endosulfan hydroxyether dehydrogenase activity
3-hydroxy-2-methylhexanoyl-CoA dehydrogenase activity
3-hydroxy-2,6-dimethyl-5-methylene-heptanoyl-CoA dehydrogenase activity
versicolorin reductase activity
ketoreductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400016151.mm.1 NM_026819.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400016151.mm.1 NM_026819.3 Refseq
Affymetrix Mouse Clariom S Array PSR1400016151.mm.1 TC1400002100.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0