This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00517333_m1
See other CRTAP GE Assays ›
Gene Symbol:
Crtap
Gene Name:
cartilage associated protein
Gene Aliases:
5730529N23Rik, CASP, Leprel3, P3h5
Chromosome Location:
Chr.9: 114375131 - 114390712 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00517333_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Crtap
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019922.2 NP_064306.2 3 - 4 898 74
GenBank mRNA AJ006469.1 - 3 - 4 856 74
AK047506.1 - 3 - 4 854 74
BC049890.1 - 3 - 4 835 6306169 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900003168.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Crtap

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s)

peptidyl-proline hydroxylation to 3-hydroxy-L-proline
protein stabilization
negative regulation of post-translational protein modification

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900026824.mm.1 NM_019922.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900026824.mm.1 NM_019922.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900026824.mm.1 TC0900003168.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0