This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00517288_g1
See other ANAPC7 GE Assays ›
Gene Symbol:
Anapc7
Gene Name:
anaphase promoting complex subunit 7
Gene Aliases:
APC7, AW545589
Chromosome Location:
Chr.5: 122422444 - 122444912 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00517288_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Anapc7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019805.4 NP_062779.3 4 - 5 649 68
GenBank mRNA AF076607.2 - 4 - 5 613 68
AK030775.1 - 4 - 5 614 68
AK049968.1 - 4 - 5 588 68
AK137286.1 - 4 - 5 613 68
AK150764.1 - 4 - 5 613 68
AK150891.1 - 4 - 5 613 68
AK153114.1 - 4 - 5 613 68
AK159523.1 - 4 - 5 581 68
AK164202.1 - 4 - 5 618 68
AK188802.1 - 1 - 2 116 68
BC006635.1 - 4 - 5 602 3482261 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001394.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Anapc7

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
cell division
protein K11-linked ubiquitination
protein phosphatase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500012338.mm.1 NM_019805.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500012338.mm.1 NM_019805.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500012338.mm.1 TC0500001394.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0