This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00517094_m1
See other RCAN1 GE Assays ›
Gene Symbol:
Rcan1
Gene Name:
regulator of calcineurin 1
Gene Aliases:
Adapt78, CALP1L, CSP1, DSC1, Dscr1, MCIP1, RCN1
Chromosome Location:
Chr.16: 92391951 - 92466169 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00517094_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rcan1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081549.2 NP_001075018.1 1 - 2 336 77
GenBank mRNA AF237789.1 - 1 - 2 86 77
AF260717.1 - 1 - 2 100 77
AF282255.1 - 1 - 2 93 77
AK146764.1 - 1 - 2 311 77
AY325904.1 - 1 - 2 292 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600002094.mm.1

Set Membership:

Amplicon length between 71 and 85 Mus musculus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

intercellular signal molecule

Gene Ontology Categories:

Function(s) Process(es)

response to ischemia
response to stress
response to oxidative stress
short-term memory
calcium-mediated signaling
locomotion involved in locomotory behavior
calcineurin-NFAT signaling cascade
regulation of phosphoprotein phosphatase activity
skeletal muscle fiber development
regulation of catalytic activity
regulation of calcineurin-NFAT signaling cascade
protein binding
calcium-dependent protein serine/threonine phosphatase regulator activity
identical protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600016107.mm.1 NM_001081549.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600016107.mm.1 TC1600002094.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 921513 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-c694fbbc8-74xkl:80/100.66.128.6:80.
git-commit: 2cf2d5f40d96ab2bfa499d940143ca22e465fdb8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0