This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00517035_m1
See other GRK5 GE Assays ›
Gene Symbol:
Grk5
Gene Name:
G protein-coupled receptor kinase 5
Gene Aliases:
Gprk5
Chromosome Location:
Chr.19: 60889593 - 61095968 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00517035_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Grk5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_018869.3 NP_061357.3 4 - 5 1173 123
GenBank mRNA AF040746.1 - 4 - 5 583 123
AK135076.1 - 4 - 5 1175 123
AK156465.1 - 4 - 5 650 123
AK161821.1 - 4 - 5 653 123
BC019379.1 - 4 - 5 446 5116510 123

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000866.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Grk5

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

desensitization of G-protein coupled receptor protein signaling pathway
protein phosphorylation
apoptotic process
signal transduction
tachykinin receptor signaling pathway
regulation of G-protein coupled receptor protein signaling pathway
positive regulation of cell proliferation
Wnt signaling pathway
phosphorylation
termination of G-protein coupled receptor signaling pathway
negative regulation of apoptotic process
protein autophosphorylation
regulation of cell cycle
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
G-protein coupled receptor kinase activity
ATP binding
lipid binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
beta-adrenergic receptor kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900007475.mm.1 NM_018869.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900007475.mm.1 NM_018869.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900007475.mm.1 TC1900000866.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0