This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00516379_g1
See other GNB3 GE Assays ›
Gene Symbol:
Gnb3
Gene Name:
guanine nucleotide binding protein (G protein), beta 3
Gene Aliases:
Gnb-3
Chromosome Location:
Chr.6: 124834240 - 124840275 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00516379_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gnb3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013530.1 NP_038558.1 1 - 2 352 89
GenBank mRNA AK014789.1 - 1 - 2 71 89
AK030354.1 - 1 - 2 35 89
BC018239.1 - 1 - 2 352 4511683 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600003121.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK014789.1,AK030354.1,BC018239.1,NM_013530.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gnb3

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
G-protein coupled receptor signaling pathway
GTPase activity
signal transducer activity
spectrin binding
GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600024300.mm.1 NM_013530.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600024300.mm.1 NM_013530.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600024300.mm.1 TC0600003121.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600024298.mm.1 NM_013530.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600024298.mm.1 NM_013530.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600024298.mm.1 TC0600003121.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600024299.mm.1 NM_013530.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600024299.mm.1 NM_013530.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600024299.mm.1 TC0600003121.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600024301.mm.1 NM_013530.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600024301.mm.1 NM_013530.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600024301.mm.1 TC0600003121.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0