This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00516033_g1
See other IDH1 GE Assays ›
Gene Symbol:
Idh1
Gene Name:
isocitrate dehydrogenase 1 (NADP+), soluble
Gene Aliases:
AI314845, AI788952, E030024J03Rik, Id-1, Idh-1, Idpc
Chromosome Location:
Chr.1: 65158616 - 65186479 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00516033_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Idh1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001111320.1 NP_001104790.1 7 - 8 1181 66
NM_010497.3 NP_034627.3 7 - 8 1159 66
GenBank mRNA AF020039.1 - 6 - 7 1009 66
AK087063.1 - 7 - 8 1061 66
AK149019.1 - 7 - 8 1090 66
AK151212.1 - 7 - 8 1084 66
AK159173.1 - 7 - 8 1090 66
AK160896.1 - 7 - 8 1091 66
AK167158.1 - 7 - 8 1131 66
AK167583.1 - 7 - 8 1095 66
AK168832.1 - 7 - 8 1132 66
BC088986.1 - 7 - 8 1131 6808731 66
DQ403075.1 - 6 - 7 909 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100002481.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Idh1

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

glyoxylate cycle
tricarboxylic acid cycle
isocitrate metabolic process
2-oxoglutarate metabolic process
glutathione metabolic process
response to oxidative stress
response to organic cyclic compound
response to steroid hormone
oxidation-reduction process
regulation of phospholipid catabolic process
regulation of phospholipid biosynthetic process
magnesium ion binding
isocitrate dehydrogenase (NADP+) activity
receptor binding
oxidoreductase activity
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
protein homodimerization activity
metal ion binding
NADP binding
NAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0