This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00515848_g1
See other GPD1 GE Assays ›
Gene Symbol:
Gpd1
Gene Name:
glycerol-3-phosphate dehydrogenase 1 (soluble)
Gene Aliases:
AI747587, Gdc-1, Gdc1, mKIAA4010
Chromosome Location:
Chr.15: 99717593 - 99725008 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00515848_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gpd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010271.2 NP_034401.1 4 - 5 515 81
GenBank mRNA AK004565.1 - 4 - 5 520 81
AK018733.1 - 4 - 5 521 81
AK137134.1 - 4 - 5 522 81
AK220445.1 - 4 - 5 509 81
BC005756.1 - 4 - 5 482 81
BC019391.1 - 4 - 5 515 4194849 81
J02655.1 - 4 - 5 507 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001023.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gpd1

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

carbohydrate metabolic process
glycerol-3-phosphate metabolic process
gluconeogenesis
NADH oxidation
glycerophosphate shuttle
NADH metabolic process
positive regulation of glycolytic process
glycerol-3-phosphate catabolic process
glycerolipid metabolic process
oxidation-reduction process
cellular response to cAMP
cellular response to tumor necrosis factor
glycerol-3-phosphate dehydrogenase [NAD+] activity
glycerol-3-phosphate dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
protein homodimerization activity
NAD binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500008079.mm.1 NM_010271.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500008079.mm.1 NM_010271.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500008079.mm.1 TC1500001023.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0