This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00515588_g1
See other CTSD GE Assays ›
Gene Symbol:
Ctsd
Gene Name:
cathepsin D
Gene Aliases:
CD, CatD
Chromosome Location:
Chr.7: 142375916 - 142387870 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00515588_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ctsd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009983.2 NP_034113.1 6 - 7 891 55
GenBank mRNA AK089294.1 - 6 - 7 849 55
AK150363.1 - 6 - 7 852 55
AK150364.1 - 6 - 7 849 55
AK150579.1 - 6 - 7 849 55
AK150622.1 - 6 - 7 849 55
AK150636.1 - 6 - 7 849 55
AK150694.1 - 6 - 7 849 55
AK150768.1 - 6 - 7 849 55
AK150946.1 - 6 - 7 901 55
AK151256.1 - 6 - 7 849 55
AK151275.1 - 6 - 7 849 55
AK151276.1 - 6 - 7 849 55
AK151374.1 - 6 - 7 849 55
AK151829.1 - 6 - 7 849 55
AK151962.1 - 6 - 7 852 55
AK152036.1 - 6 - 7 862 55
AK152045.1 - 6 - 7 902 55
AK152168.1 - 6 - 7 849 55
AK152187.1 - 6 - 7 849 55
AK152188.1 - 6 - 7 849 55
AK152406.1 - 6 - 7 852 55
AK152458.1 - 6 - 7 849 55
AK152567.1 - 6 - 7 849 55
AK152578.1 - 6 - 7 849 55
AK152593.1 - 6 - 7 879 55
AK152680.1 - 6 - 7 849 55
AK152828.1 - 6 - 7 849 55
AK152881.1 - 6 - 7 849 55
AK152892.1 - 6 - 7 849 55
AK153191.1 - 6 - 7 849 55
AK153291.1 - 6 - 7 849 55
AK159097.1 - 6 - 7 849 55
AK159440.1 - 6 - 7 902 55
AK159490.1 - 6 - 7 849 55
AK159512.1 - 6 - 7 891 55
AK159580.1 - 6 - 7 849 55
AK159642.1 - 4 - 5 601 55
AK159743.1 - 6 - 7 853 55
AK159896.1 - 6 - 7 849 55
AK159970.1 - 6 - 7 902 55
AK165227.1 - 6 - 7 919 55
AK166864.1 - 6 - 7 862 55
AK166938.1 - 6 - 7 896 55
AK167143.1 - 6 - 7 887 55
AK167366.1 - 6 - 7 921 55
AK167698.1 - 6 - 7 848 55
AK167750.1 - 6 - 7 862 55
AK167867.1 - 6 - 7 850 55
AK169139.1 - 6 - 7 902 55
AK171195.1 - 6 - 7 850 55
AK172583.1 - 6 - 7 850 55
AK213803.1 - 1 - 2 43 55
BC019682.1 - 2 - 3 246 55
BC054758.1 - 6 - 7 921 6413315 55
BC057931.1 - 6 - 7 882 5253319 55
X52886.1 - 6 - 7 891 55
X53337.1 - 6 - 7 831 55

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004565.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ctsd

Gene Ontology Categories:

Function(s) Process(es)

autophagosome assembly
proteolysis
autophagy
protein catabolic process
epithelial tube branching involved in lung morphogenesis
endopeptidase activity
aspartic-type endopeptidase activity
peptidase activity
hydrolase activity
peptide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0