This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00515579_m1
See other CTSC GE Assays ›
Gene Symbol:
Ctsc
Gene Name:
cathepsin C
Gene Aliases:
AI047818, DPP1, DPPI
Chromosome Location:
Chr.7: 88278023 - 88310875 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00515579_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ctsc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009982.4 NP_034112.3 3 - 4 594 97
GenBank mRNA AK002454.1 - 3 - 4 553 97
AK049406.1 - 3 - 4 553 97
AK146858.1 - 3 - 4 602 97
AK150761.1 - 3 - 4 554 97
AK151711.1 - 3 - 4 554 97
AK151861.1 - 3 - 4 440 97
AK153168.1 - 3 - 4 554 97
AK167882.1 - 3 - 4 553 97
BC067063.1 - 3 - 4 527 30541061 97
U74683.3 - 3 - 4 551 97
U89269.1 - 3 - 4 552 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001258.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ctsc

Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

T cell mediated cytotoxicity
proteolysis
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
response to organic substance
positive regulation of peptidase activity
proteolysis involved in cellular protein catabolic process
positive regulation of proteolysis involved in cellular protein catabolic process
positive regulation of apoptotic signaling pathway
cysteine-type endopeptidase activity
serine-type endopeptidase activity
peptidase activity
cysteine-type peptidase activity
cysteine-type endopeptidase activator activity involved in apoptotic process
peptidase activator activity involved in apoptotic process
hydrolase activity
phosphatase binding
chloride ion binding
identical protein binding
protein self-association
chaperone binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700010880.mm.1 NM_009982.5 Refseq
Affymetrix Mouse Clariom S Array PSR0700010880.mm.1 TC0700001258.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0