This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00515309_m1
See other DPYSL3 GE Assays ›
Gene Symbol:
Dpysl3
Gene Name:
dihydropyrimidinase-like 3
Gene Aliases:
CRMP-4, DRP-3, TUC4, ULIP-1, Ulip, Ulip1
Chromosome Location:
Chr.18: 43320979 - 43438286 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00515309_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dpysl3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001136086.2 NP_001129558.1 3 - 4 474 86
NM_001291455.1 NP_001278384.1 3 - 4 724 86
NM_009468.5 NP_033494.1 3 - 4 655 86
GenBank mRNA AK077000.1 - 3 - 4 465 86
AK133040.1 - 3 - 4 474 86
AK149024.1 - 3 - 4 493 86
AK161509.1 - 3 - 4 396 86
BC023003.1 - 3 - 4 339 5367381 86
X87817.1 - 3 - 4 615 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1800001269.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dpysl3

Panther Classification:

Molecular Function -

hydrolase

Gene Ontology Categories:

Function(s) Process(es)

nervous system development
positive regulation of neuron projection development
negative regulation of neuron projection development
negative regulation of cell migration
actin filament bundle assembly
protein homooligomerization
positive regulation of filopodium assembly
actin crosslink formation
protein binding
hydrolase activity
hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds
SH3 domain binding
chondroitin sulfate binding
phosphoprotein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1800009445.mm.1 NM_001136086.2 Refseq
Affymetrix Mouse Clariom S Array PSR1800009445.mm.1 NM_009468.5 Refseq
Affymetrix Mouse Clariom S Array PSR1800009445.mm.1 TC1800001269.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0