This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00515240_g1
See other KIF2A GE Assays ›
Gene Symbol:
Kif2a
Gene Name:
kinesin family member 2A
Gene Aliases:
C530030B14Rik, Kif2, Kns2, M-kinesin
Chromosome Location:
Chr.13: 106960585 - 107022228 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00515240_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Kif2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145779.1 NP_001139251.1 10 - 11 999 102
NM_008442.2 NP_032468.2 11 - 12 1382 102
GenBank mRNA AK037037.1 - 10 - 11 912 102
BC006803.1 - 10 - 11 994 3152883 102
D12644.1 - 11 - 12 1383 102
Y15894.1 - 9 - 10 849 102

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300002619.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Kif2a

Panther Classification:

Molecular Function -

microtubule binding motor protein

Gene Ontology Categories:

Function(s) Process(es)

microtubule-based movement
microtubule depolymerization
cell cycle
mitotic spindle organization
mitotic nuclear division
multicellular organism development
nervous system development
cell differentiation
cell division
mitotic spindle assembly
nucleotide binding
microtubule motor activity
ATP binding
microtubule binding
ATPase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300017847.mm.1 NM_001145779.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017847.mm.1 NM_001145779.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017847.mm.1 NM_008442.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300017847.mm.1 NM_008442.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300017847.mm.1 TC1300002619.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0