This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00515156_m1
See other IGFBP3 GE Assays ›
Gene Symbol:
Igfbp3
Gene Name:
insulin-like growth factor binding protein 3
Gene Aliases:
AI649005, IGFBP-3, IGgfbp3
Chromosome Location:
Chr.11: 7206086 - 7213931 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00515156_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igfbp3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008343.2 NP_032369.2 1 - 2 537 66
GenBank mRNA AK077477.1 - 1 - 2 78 66
AK141690.1 - 1 - 2 159 66
BC031421.1 - 1 - 2 496 66
BC058261.1 - 1 - 2 507 6487655 66
X81581.1 - 1 - 2 485 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002195.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Igfbp3

Panther Classification:

Molecular Function -

protease inhibitor

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell growth
negative regulation of protein phosphorylation
protein phosphorylation
negative regulation of cell proliferation
regulation of glucose metabolic process
negative regulation of smooth muscle cell migration
regulation of growth
positive regulation of apoptotic process
positive regulation of catalytic activity
positive regulation of MAPK cascade
positive regulation of insulin-like growth factor receptor signaling pathway
type B pancreatic cell proliferation
positive regulation of myoblast differentiation
negative regulation of smooth muscle cell proliferation
fibronectin binding
insulin-like growth factor binding
protein tyrosine phosphatase activator activity
growth factor binding
insulin-like growth factor I binding
insulin-like growth factor II binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0