This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00515147_g1
See other IDUA GE Assays ›
Gene Symbol:
Idua
Gene Name:
iduronidase, alpha-L-
Gene Aliases:
6030426D08
Chromosome Location:
Chr.5: 108660331 - 108685446 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00515147_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Idua
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008325.4 NP_032351.2 8 - 9 1395 75
NM_172997.1 NP_766585.1 7 - 8 1254 75
GenBank mRNA AK031414.1 - 7 - 8 1049 75
AK045330.1 - 6 - 7 882 75
AK155627.1 - 8 - 9 1396 75
AK169998.1 - 7 - 8 1053 75
AK171124.1 - 3 - 4 406 75
BC057935.1 - 7 - 8 1016 5327150 75
L34111.1 - 7 - 8 969 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001112.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Idua

Panther Classification:

Molecular Function -

glycosidase

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
carbohydrate metabolic process
lysosome organization
metabolic process
dermatan sulfate catabolic process
limb morphogenesis
skeletal system morphogenesis
chemical homeostasis
L-iduronidase activity
hydrolase activity, hydrolyzing O-glycosyl compounds
hydrolase activity
hydrolase activity, acting on glycosyl bonds

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500009493.mm.1 NM_008325.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500009493.mm.1 NM_172997.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500009493.mm.1 TC0500001112.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0