Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700038967.mm.1 |
NM_001252572.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038967.mm.1 |
NM_001252572.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038967.mm.1 |
NM_007803.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038967.mm.1 |
NM_007803.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038967.mm.1 |
TC0700004604.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700038965.mm.1 |
NM_001252572.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038965.mm.1 |
NM_001252572.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038965.mm.1 |
NM_007803.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038965.mm.1 |
NM_007803.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038965.mm.1 |
TC0700004604.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700038964.mm.1 |
NM_001252572.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038964.mm.1 |
NM_001252572.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038964.mm.1 |
NM_007803.5.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038964.mm.1 |
NM_007803.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700038964.mm.1 |
TC0700004604.mm.1 |
MTA TC |