This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00513595_m1
See other SPON2 GE Assays ›
Gene Symbol:
Spon2
Gene Name:
spondin 2, extracellular matrix protein
Gene Aliases:
2310045I24Rik, AI504350, M-spondin, Mindin, Mspondin
Chromosome Location:
Chr.5: 33213518 - 33218253 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00513595_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Spon2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133903.3 NP_598664.3 4 - 5 920 64
GenBank mRNA AK028987.1 - 4 - 5 865 64
AK042840.1 - 3 - 4 940 64
AK131131.1 - 3 - 4 503 64
AY457639.1 - 4 - 5 655 64
BC017616.1 - 4 - 5 853 4923292 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002213.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Spon2

Panther Classification:

Molecular Function -

cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

immune system process
mast cell mediated immunity
cell adhesion
opsonization
response to lipopolysaccharide
positive regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
defense response to bacterium
induction of bacterial agglutination
innate immune response
defense response to fungus
defense response to virus
positive regulation of macrophage cytokine production
cellular response to lipopolysaccharide
lipopolysaccharide binding
antigen binding
receptor binding
protein binding
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500020165.mm.1 NM_133903.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500020165.mm.1 NM_133903.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500020165.mm.1 TC0500002213.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 917195 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0