This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00513038_m1
See other MTFR1L GE Assays ›
Gene Symbol:
Mtfr1l
Gene Name:
mitochondrial fission regulator 1-like
Gene Aliases:
2410166I05Rik, AI480750, Fam54b
Chromosome Location:
Chr.4: 134525555 - 134535387 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00513038_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mtfr1l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256112.1 NP_001243041.1 3 - 4 424 152
NM_029759.4 NP_084035.1 3 - 4 375 152
GenBank mRNA AK010825.1 - 3 - 4 416 152
AK080126.1 - 2 - 3 2744 152
AK089141.1 - 3 - 4 416 152
AK145362.1 - 3 - 4 416 152
AK146416.1 - 2 - 3 251 152
AK164004.1 - 2 - 3 251 152
AK169047.1 - 3 - 4 283 152
AK171703.1 - 3 - 4 416 152
AK187934.1 - 3 - 4 256 153
AK216840.1 - 2 - 3 241 152
BC026808.1 - 2 - 3 247 4159289 152

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003740.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mtfr1l

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial fission
biological_process
aerobic respiration
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400030791.mm.1 NM_001256112.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030791.mm.1 NM_001256112.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030791.mm.1 NM_029759.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030791.mm.1 NM_029759.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400030791.mm.1 TC0400003740.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0