This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00511963_m1
See other ACOXL GE Assays ›
Gene Symbol:
Acoxl
Gene Name:
acyl-Coenzyme A oxidase-like
Gene Aliases:
1200014P05Rik, AV025673
Chromosome Location:
Chr.2: 127838797 - 128126252 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00511963_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acoxl
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028765.3 NP_083041.1 1 - 2 357 81
GenBank mRNA AK004775.1 - 1 - 2 359 81

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acoxl

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

fatty acid metabolic process
fatty acid beta-oxidation
metabolic process
oxidation-reduction process
acyl-CoA dehydrogenase activity
acyl-CoA oxidase activity
palmitoyl-CoA oxidase activity
pristanoyl-CoA oxidase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors
very-long-chain fatty acyl-CoA oxidase activity
flavin adenine dinucleotide binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-9c87ff496-8j9ll:80/100.66.128.142:80.
git-commit: 146a63324c51a096ecccde4707f5fe4015d9ef66
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36_secrets_rm