This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00511025_g1
See other CPSF3L GE Assays ›
Gene Symbol:
Cpsf3l
Gene Name:
cleavage and polyadenylation specific factor 3-like
Gene Aliases:
2410006F12Rik, AU044035
Chromosome Location:
Chr.4: 155869548 - 155889103 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00511025_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cpsf3l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028020.3 NP_082296.1 15 - 16 1728 75
GenBank mRNA AK010425.1 - 15 - 16 1690 75
AK090206.1 - 15 - 16 1729 75
AK150436.1 - 15 - 16 1725 75
AK152740.1 - 15 - 16 1725 75
AK172533.1 - 15 - 16 1712 75
BC008240.1 - 7 - 8 824 3601302 75
BC011155.1 - 15 - 16 1712 4195656 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002097.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cpsf3l

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

snRNA processing
molecular_function
hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400017574.mm.1 NM_028020.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017574.mm.1 NM_028020.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400017574.mm.1 TC0400002097.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400017576.mm.1 NM_028020.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017576.mm.1 NM_028020.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400017576.mm.1 TC0400002097.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0