This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00510996_g1
See other CERS5 GE Assays ›
Gene Symbol:
Cers5
Gene Name:
ceramide synthase 5
Gene Aliases:
2310081H14Rik, AU045339, AW544927, Lass5, Trh4
Chromosome Location:
Chr.15: 99735592 - 99772515 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00510996_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cers5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_028015.2 NP_082291.1 7 - 8 907 61
GenBank mRNA AK010241.1 - 7 - 8 835 61
AK075694.1 - 7 - 8 827 61
AK145589.1 - 7 - 8 815 61
AK145762.1 - 7 - 8 829 61
AK146220.1 - 7 - 8 829 61
AK146417.1 - 7 - 8 799 61
AY029533.1 - 7 - 8 827 61
BC010670.1 - 1 - 2 128 61
BC043059.1 - 854 6419704 61
BC046797.1 - 7 - 8 907 6403129 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500002221.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cers5

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
sphingolipid metabolic process
sphingolipid biosynthetic process
ceramide biosynthetic process
DNA binding
transferase activity
sphingosine N-acyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500018219.mm.1 NM_028015.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500018219.mm.1 NM_028015.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500018219.mm.1 TC1500002221.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0