This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00509987_m1
See other ERP27 GE Assays ›
Gene Symbol:
Erp27
Gene Name:
endoplasmic reticulum protein 27
Gene Aliases:
1810033M07Rik, 1810047B09Rik
Chromosome Location:
Chr.6: 136907314 - 136922461 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00509987_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Erp27
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026983.2 NP_081259.1 4 - 5 510 129
GenBank mRNA AK007684.1 - 4 - 5 511 129
AK131830.1 - 4 - 5 511 129
BC037376.1 - 3 - 4 346 129
BC104406.1 - 4 - 5 40050532 129
BC104407.2 - 4 - 5 40050533 129

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600003336.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Erp27

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094292 AK131830.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094292 NM_026983.2 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094292 AK007684.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 636815 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 787809 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481632 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 672865 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 147202 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 151275 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 971394 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970177 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 682765 AK032011.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 922226 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 78210 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172278 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179699 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094292 AK131830.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094292 NM_026983.2 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2094292 AK007684.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875222 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875212 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875217 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875231 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875232 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875218 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875234 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875227 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875237 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875236 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875230 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875215 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875214 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875216 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875221 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875219 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875210 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875229 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875223 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24875211 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179699 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172278 - -
Whole Mouse Genome G4122A A_52_P366696 BC037376.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P366696 BC104406.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P366696 BC104407.2 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P366696 AK131830.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P366696 NM_026983.2 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172278 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179699 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05776663 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05776660 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05523956 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05800013 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875156 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875160 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875176 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875210 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875143 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875198 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875170 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875182 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875138 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875219 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875223 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875231 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875175 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875218 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875091 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875123 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875196 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875161 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875105 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875173 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875136 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875181 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875153 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875165 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875098 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875221 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875132 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875128 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875093 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875185 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875097 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875112 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875151 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875134 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875167 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875205 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875163 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875212 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875090 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875222 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875187 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875140 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875190 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875127 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875121 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875118 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875229 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875158 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875236 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875211 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875114 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875147 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875206 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875192 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875227 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875177 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875131 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875237 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875092 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875159 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875149 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875145 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875110 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875096 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875193 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875200 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875201 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875214 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875183 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875120 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875141 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875171 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875135 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875197 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875216 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875094 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24875146 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05435845 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875197 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875194 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05006773 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875180 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875134 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875196 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875195 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875222 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05006764 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875198 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875182 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875232 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05024255 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875218 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875201 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875219 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875223 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875200 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05180423 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875192 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875183 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05024254 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875205 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05342670 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875193 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875236 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875136 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875143 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05006768 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875141 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875231 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875181 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875206 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875234 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875221 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875237 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875202 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875229 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875210 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875227 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875185 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875187 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875135 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875179 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875208 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24875178 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10548888 - -
Affymetrix Mouse Gene 1.0 ST Array 10548887 - -
Affymetrix Mouse Gene 1.0 ST Array 10548890 - -
Affymetrix Mouse Gene 1.0 ST Array 10548891 - -
Affymetrix Mouse Gene 1.0 ST Array 10548885 - -
Affymetrix Mouse Gene 1.0 ST Array 10548889 - -
Affymetrix Mouse Gene 1.0 ST Array 10548886 - -
Affymetrix HT MG-430 PM Array Plate 1430913_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1452695_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10548884 - -
Mouse Genome 430 2.0 Array 1452695_at - -
Mouse Genome 430 2.0 Array 1430913_at - -
Mouse Genome 430 2.0 Array 1430913_PM_at - -
Mouse Genome 430 2.0 Array 1452695_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 69187_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 69187_at - -
Affymetrix Mouse Clariom S Array PSR0600026180.mm.1 NM_026983.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600026180.mm.1 NM_026983.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600026180.mm.1 TC0600003336.mm.1 MTA TC
Mouse Expression Set 430 1452695_at - -
Mouse Expression Set 430 1452695_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1452695_at - -
Affymetrix HT Mouse Genome 430B Array 1452695_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1430913_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1430913_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 418903 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 6380142 - -
Illumina mouse-6 v1.0 expression beadchip 6380142 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006380142 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2658990 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2658990 - -
mouse-6 v1.1 (Illumina) scl28273.7.1_5-S - -
Illumina Sentrix mouse-6.v1 bead array 6380142 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl28273.7.1_5-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6380142 - -
MouseRef-8 v2 Expression BeadChip ILMN_2888940 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2888940 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl28273.7.1_5-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0