This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00509522_m1
See other S100A16 GE Assays ›
Gene Symbol:
S100a16
Gene Name:
S100 calcium binding protein A16
Gene Aliases:
2300002L21Rik, AI325039, AI663996, DT1P1A7, S100F
Chromosome Location:
Chr.3: 90537257 - 90543151 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00509522_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: S100a16
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK009762.1 - 1 - 2 47 71
AK160178.1 - 1 - 2 47 71
AK160512.1 - 1 - 2 39 71
AK167006.1 - 1 - 2 39 71

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK009762.1, AK160178.1, AK160512.1, AK167006.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mus musculus 1700 Inventoried Probe spans exons

Panther Classification:

Molecular Function -

calmodulin-related

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 917377 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0