This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00509158_g1
See other EPS8L1 GE Assays ›
Gene Symbol:
Eps8l1
Gene Name:
EPS8-like 1
Gene Aliases:
2310051G19Rik, 4632407K17Rik, AW060268, DRC3, EPS8R1
Chromosome Location:
Chr.7: 4460631 - 4480487 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00509158_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eps8l1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290416.1 NP_001277345.1 11 - 12 1218 57
NM_026146.4 NP_080422.1 11 - 12 1124 57
GenBank mRNA AK014556.1 - 11 - 12 1219 57
AK019490.2 - 11 - 12 1118 57
AY074931.1 - 10 - 11 1061 57
BC027043.1 - 11 - 12 1408 57
BC100383.1 - 11 - 12 1306 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000033.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eps8l1

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

Rho protein signal transduction
Rac protein signal transduction
regulation of Rho protein signal transduction
positive regulation of GTPase activity
positive regulation of ruffle assembly
actin binding
Rho guanyl-nucleotide exchange factor activity
Rac guanyl-nucleotide exchange factor activity
T cell receptor binding
actin filament binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700000445.mm.1 NM_026146.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700000445.mm.1 TC0700000033.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700000446.mm.1 NM_026146.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700000446.mm.1 TC0700000033.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0