This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00508496_m1
See other CMSS1 GE Assays ›
Gene Symbol:
Cmss1
Gene Name:
cms small ribosomal subunit 1
Gene Aliases:
1110001A06Rik, 2610528E23Rik, 4930572F24Rik, AV099812, AV122629
Chromosome Location:
Chr.16: 57302000 - 57606867 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00508496_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cmss1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025599.3 NP_079875.2 5 - 6 554 83
GenBank mRNA AK012168.1 - 5 - 6 535 83
AK019979.1 - 5 - 6 530 83
BC038891.1 - 5 - 6 441 3371329 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600001761.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cmss1

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600014322.mm.1 NM_025599.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600014322.mm.1 NM_025599.3 Refseq
Affymetrix Mouse Clariom S Array PSR1600014322.mm.1 TC1600001761.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0