This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00508063_m1
See other ANAPC5 GE Assays ›
Gene Symbol:
Anapc5
Gene Name:
anaphase-promoting complex subunit 5
Gene Aliases:
2510006G12Rik, AA408751, AA536819, AA986414, APC5, Anpc5
Chromosome Location:
Chr.5: 122787459 - 122821342 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00508063_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Anapc5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042491.2 NP_001035956.1 2 - 3 436 61
NM_001289517.1 NP_001276446.1 2 - 3 436 61
NM_001289518.1 NP_001276447.1 2 - 3 436 61
NM_001289519.1 NP_001276448.1 2 - 3 436 61
NM_001289520.1 NP_001276449.1 2 - 3 436 61
NM_021505.3 NP_067480.2 2 - 3 436 61
GenBank mRNA AB041593.1 - 2 - 3 379 61
AK013978.1 - 2 - 3 360 61
AK031432.1 - 2 - 3 361 61
AK075969.1 - 2 - 3 375 61
AK088327.1 - 2 - 3 399 61
AK147951.1 - 2 - 3 375 61
AK151314.1 - 2 - 3 382 61
AK151905.1 - 2 - 3 382 61
AK152981.1 - 2 - 3 382 61
AK153373.1 - 2 - 3 382 61
AK153422.1 - 2 - 3 382 61
AK159204.1 - 2 - 3 382 61
AK159713.1 - 2 - 3 375 61
AK159916.1 - 2 - 3 375 61
AK167561.1 - 2 - 3 375 61
AK168135.1 - 2 - 3 360 61
AK168737.1 - 2 - 3 376 61
AK168814.1 - 2 - 3 375 61
AK169196.1 - 2 - 3 399 61
AK180837.1 - 2 - 3 109 61
AK192754.1 - 2 - 3 132 61
AK194108.1 - 2 - 3 101 61
AK197886.1 - 2 - 3 108 61
AK203216.1 - 1 - 2 85 61
AK215079.1 - 2 - 3 109 61
AK217317.1 - 2 - 3 114 61
BC010339.1 - 2 - 3 352 2648173 61
BC046566.1 - 2 - 3 415 2650790 61
BC046804.1 - 2 - 3 373 6404923 61
BQ950394.1 - 2 - 3 436 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003220.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Anapc5

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
mitotic nuclear division
cell division
protein K11-linked ubiquitination
protein phosphatase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500028818.mm.1 NM_001042491.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500028818.mm.1 NM_001289517.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500028818.mm.1 NM_001289518.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500028818.mm.1 NM_001289519.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500028818.mm.1 NM_001289520.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500028818.mm.1 NM_021505.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500028818.mm.1 TC0500003220.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0