Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0300005860.mm.1 |
NM_001038587.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005860.mm.1 |
NM_001038587.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005860.mm.1 |
NM_001146296.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005860.mm.1 |
NM_001146296.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005860.mm.1 |
NM_019655.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005860.mm.1 |
NM_019655.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005860.mm.1 |
TC0300000769.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300005857.mm.1 |
NM_001146296.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005857.mm.1 |
NM_001146296.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005857.mm.1 |
NM_019655.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005857.mm.1 |
NM_019655.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005857.mm.1 |
TC0300000769.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300005858.mm.1 |
NM_001146296.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005858.mm.1 |
NM_001146296.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005858.mm.1 |
NM_019655.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005858.mm.1 |
NM_019655.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005858.mm.1 |
TC0300000769.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300005859.mm.1 |
NM_001038587.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005859.mm.1 |
NM_001038587.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005859.mm.1 |
NM_001146296.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005859.mm.1 |
NM_001146296.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005859.mm.1 |
NM_019655.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005859.mm.1 |
NM_019655.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005859.mm.1 |
TC0300000769.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0300005856.mm.1 |
NM_001146296.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005856.mm.1 |
NM_001146296.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005856.mm.1 |
NM_019655.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005856.mm.1 |
NM_019655.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0300005856.mm.1 |
TC0300000769.mm.1 |
MTA TC |