This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00507950_m1
See other ADAT1 GE Assays ›
Gene Symbol:
Adat1
Gene Name:
adenosine deaminase, tRNA-specific 1
Gene Aliases:
MMADAT1, mADAT1
Chromosome Location:
Chr.8: 111966897 - 111992307 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00507950_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adat1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013925.4 NP_038953.1 5 - 6 583 77
GenBank mRNA AF192375.1 - 4 - 5 422 77
AK035614.1 - 5 - 6 571 77
AK039699.1 - 5 - 6 553 77
AK082399.1 - 5 - 6 584 77
AK161044.1 - 5 - 6 553 77
BC019976.1 - 5 - 6 529 4458235 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002960.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adat1

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

RNA processing
tRNA processing
RNA binding
adenosine deaminase activity
tRNA-specific adenosine deaminase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800022871.mm.1 NM_013925.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022871.mm.1 NM_013925.4 Refseq
Affymetrix Mouse Clariom S Array PSR0800022871.mm.1 TC0800002960.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0