Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400003294.mm.1 |
NM_001281845.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003294.mm.1 |
NM_013473.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003294.mm.1 |
TC1400000368.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003293.mm.1 |
NM_001281845.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003293.mm.1 |
NM_013473.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003293.mm.1 |
TC1400000368.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003296.mm.1 |
NM_001281845.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003296.mm.1 |
NM_013473.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003296.mm.1 |
TC1400000368.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003298.mm.1 |
NM_001281845.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003298.mm.1 |
NM_013473.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003298.mm.1 |
TC1400000368.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003292.mm.1 |
NM_013473.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003292.mm.1 |
TC1400000368.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400003295.mm.1 |
NM_001281845.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003295.mm.1 |
NM_013473.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400003295.mm.1 |
TC1400000368.mm.1 |
MTA TC |