This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00507847_m1
See other AES GE Assays ›
Gene Symbol:
Aes
Gene Name:
amino-terminal enhancer of split
Gene Aliases:
AL024115, Esp1, Grg, Grg-5, Grg5, Tle5
Chromosome Location:
Chr.10: 81559444 - 81566371 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00507847_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aes
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001276288.1 NP_001263217.1 1 - 2 225 76
NM_010347.4 NP_034477.1 1 - 2 225 76
NR_074087.1 1 - 2 225 76
GenBank mRNA AK131845.1 - 1 - 2 102 76
AK131994.1 - 1 - 2 173 76
AK158714.1 - 1 - 2 110 76
BC132217.1 - 1 - 2 113 40130494 76
BC132219.1 - 1 - 2 113 40130496 76
BC145299.1 - 1 - 2 113 9053853 76
BY713872.1 - 1 - 2 226 76
L12140.1 - 1 - 2 66 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000903.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK131845.1,AK158714.1,BC132217.1,BC132219.1,BC145299.1,L12140.1,NM_001276288.1,NM_010347.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aes

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
skeletal system development
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
negative regulation of gene expression
Wnt signaling pathway
cellular response to extracellular stimulus
negative regulation of protein binding
regulation of growth
negative regulation of transcription, DNA-templated
negative regulation of response to cytokine stimulus
response to interleukin-1
negative regulation of canonical Wnt signaling pathway
positive regulation of anoikis
transcription corepressor activity
protein binding

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043287 X73359.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043287 X73361.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043287 L12140.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043287 AK149015.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2043287 AK158714.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968285 X73359.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968285 L12140.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968285 AK149015.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968285 AK158714.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1968285 NM_010347.3 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 37646 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 790022 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 902519 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 440397 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 140249 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 121047 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P162166 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043287 X73359.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043287 X73361.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043287 L12140.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043287 AK149015.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2043287 AK158714.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968285 X73359.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968285 L12140.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968285 AK149015.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968285 AK158714.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1968285 NM_010347.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243342 X73359.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243342 X73361.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243342 L12140.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243342 AK149015.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P243342 AK158714.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290505 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290509 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290500 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290490 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290508 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290481 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290497 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290503 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290491 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290482 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290512 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290494 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290477 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290484 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290486 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290498 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290511 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290483 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290485 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290493 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290488 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27290480 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07126010 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P162166 - -
Whole Mouse Genome G4122A A_51_P243342 X73359.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P243342 X73361.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P243342 L12140.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P243342 AK149015.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P243342 AK158714.1 mouse_transcriptome
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P162166 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08826767 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08374700 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290612 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290524 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290519 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290529 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290528 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290450 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290581 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290589 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290518 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290569 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290609 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290597 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290545 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290516 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290480 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290604 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290520 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290477 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290587 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290577 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290536 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290440 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290453 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290443 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290494 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290446 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290458 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290592 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290514 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290566 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290474 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290449 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290567 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290533 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290472 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290490 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290615 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290575 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290578 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290521 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290585 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290508 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290461 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290583 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290503 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290617 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290561 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290498 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290564 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290515 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290531 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290573 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290509 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290513 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290467 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290451 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290584 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290484 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290483 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290500 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290512 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290497 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290606 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290596 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290468 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290456 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290613 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290441 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290496 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290522 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290543 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290462 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290491 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290470 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290482 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290538 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290486 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290505 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27290565 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08533441 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290485 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08910676 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290508 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08582103 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08910675 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08533438 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290533 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290538 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290498 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290528 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290529 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290525 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08703829 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08943315 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290486 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290536 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290520 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08592857 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290513 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290505 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08933338 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290477 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290475 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290468 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290543 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08910660 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290491 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08826767 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290460 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290497 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290470 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08847748 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290496 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08327250 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290490 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08634394 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08533439 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290539 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290480 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08378400 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290503 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290500 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08910659 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08933339 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290461 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290462 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290518 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290474 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290476 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08879482 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290489 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290511 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27290465 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10365144 - -
Affymetrix Mouse Gene 1.0 ST Array 10365137 - -
Affymetrix Mouse Gene 1.0 ST Array 10365138 - -
Affymetrix Mouse Gene 1.0 ST Array 10365140 - -
Affymetrix Mouse Gene 1.0 ST Array 10365143 - -
Affymetrix Mouse Gene 1.0 ST Array 10365142 - -
Affymetrix Mouse Gene 1.0 ST Array 10365139 - -
Affymetrix Mouse Gene 1.0 ST Array 10365141 - -
Affymetrix HT MG-430 PM Array Plate 1420619_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10365136 - -
Mouse Genome 430 2.0 Array 1420619_PM_a_at - -
Mouse Genome 430 2.0 Array 1420619_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 14797_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 14797_at - -
Affymetrix Murine Genome 6500A Array Msa1040_at - -
Affymetrix Mouse Clariom S Array PSR1000007134.mm.1 NM_001276288.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000007134.mm.1 NM_010347.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000007134.mm.1 NR_074087.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000007134.mm.1 TC1000000903.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000007132.mm.1 NM_001276288.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000007132.mm.1 NM_010347.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000007132.mm.1 NR_074087.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000007132.mm.1 TC1000000903.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000007130.mm.1 NM_001276288.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000007130.mm.1 NM_010347.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000007130.mm.1 NR_074087.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000007130.mm.1 TC1000000903.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000007129.mm.1 NM_001276288.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000007129.mm.1 NM_010347.4 Refseq
Affymetrix Mouse Clariom S Array PSR1000007129.mm.1 NR_074087.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000007129.mm.1 TC1000000903.mm.1 MTA TC
Mouse Expression Set 430 1420619_a_at - -
Mouse Expression Set 430 1420619_PM_a_at - -
Murine Genome U74v2 Set 160215_PM_at - -
Murine Genome U74v2 Set 160215_at - -
Mouse Genome 430A 2.0 Array 1420619_PM_a_at - -
Mouse Genome 430A 2.0 Array 1420619_a_at - -
Affymetrix HT Mouse Genome 430A Array 1420619_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1420619_a_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 758404 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 6550333 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006550333 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1213591 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1213591 - -
mouse-6 v1.1 (Illumina) scl38644.7_75-S - -
Illumina Sentrix mouse-6.v1 bead array 6550333 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl38644.7_75-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6550333 - -
MouseRef-8 v2 Expression BeadChip ILMN_2808336 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2808336 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1213591 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl38644.7_75-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0