This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00507642_m1
See other SLC9A8 GE Assays ›
Gene Symbol:
Slc9a8
Gene Name:
solute carrier family 9 (sodium/hydrogen exchanger), member 8
Gene Aliases:
1200006P13Rik, 6430709P13Rik, AI182282, NHE-8, NHE8
Chromosome Location:
Chr.2: 167420056 - 167477000 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00507642_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc9a8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AB089793.1 - 7 - 8 591 82
AF482993.1 - 6 - 7 555 82
AK049410.1 - 7 - 8 587 82
AK051791.1 - 6 - 7 506 82
AK143281.1 - 7 - 8 560 82
AK173069.1 - 7 - 8 927 82
BC030879.1 - 7 - 8 590 4241988 82
BC058947.1 - 7 - 8 561 5702781 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002587.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200021319.mm.1 NM_148929.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200021319.mm.1 TC0200002587.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0