This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00506218_m1
See other ALDH6A1 GE Assays ›
Gene Symbol:
Aldh6a1
Gene Name:
aldehyde dehydrogenase family 6, subfamily A1
Gene Aliases:
1110038I05Rik, AI314632, Mmsdh
Chromosome Location:
Chr.12: 84430717 - 84450950 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00506218_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aldh6a1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134042.2 NP_598803.1 2 - 3 119 111
GenBank mRNA AF297860.1 - 2 - 3 119 111
AK033587.1 - 2 - 3 146 111
AK082790.1 - 2 - 3 173 111
AK128924.1 - 2 - 3 151 111
AK147146.1 - 2 - 3 173 111
AK155814.1 - 2 - 3 170 111
AK169915.1 - 2 - 3 159 111
AK170305.1 - 2 - 3 172 111
AK171581.1 - 2 - 3 173 111
AK171896.1 - 2 - 3 162 111
BC031148.1 - 2 - 3 244 4975560 111
BC033440.1 - 1 - 2 142 4212084 111
BC115855.1 - 2 - 3 94 111

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC031148.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Aldh6a1

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

thymine catabolic process
valine metabolic process
valine catabolic process
metabolic process
beta-alanine catabolic process
thymine metabolic process
brown fat cell differentiation
oxidation-reduction process
aldehyde dehydrogenase (NAD) activity
methylmalonate-semialdehyde dehydrogenase (acylating) activity
oxidoreductase activity
oxidoreductase activity, acting on the aldehyde or oxo group of donors, NAD or NADP as acceptor
thiolester hydrolase activity
malonate-semialdehyde dehydrogenase (acetylating) activity
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0