This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00505743_g1
See other PAPD4 GE Assays ›
Gene Symbol:
Papd4
Gene Name:
PAP associated domain containing 4
Gene Aliases:
8030446C20Rik, AI649009, GLD-2, Gld2
Chromosome Location:
Chr.13: 93146275 - 93192970 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00505743_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Papd4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133905.2 NP_598666.1 13 - 14 1522 75
GenBank mRNA AK033141.1 - 10 - 11 857 75
AK146086.1 - 13 - 14 1442 75
AM419011.1 - 12 - 13 1283 75
BC016629.1 - 13 - 14 1522 3595512 75

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Papd4

Panther Classification:

Molecular Function -

nucleotidyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
mRNA processing
RNA polyadenylation
histone mRNA catabolic process
nucleotide binding
polynucleotide adenylyltransferase activity
protein binding
ATP binding
transferase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0