This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00505051_m1
See other ATP6V1C2 GE Assays ›
Gene Symbol:
Atp6v1c2
Gene Name:
ATPase, H+ transporting, lysosomal V1 subunit C2
Gene Aliases:
1110038G14Rik
Chromosome Location:
Chr.12: 17284719 - 17325371 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00505051_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Atp6v1c2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159632.1 NP_001153104.1 12 - 13 1141 75
NM_133699.2 NP_598460.1 11 - 12 1111 75
GenBank mRNA AB088357.1 - 11 - 12 1096 75
BC056636.1 - 12 - 13 1114 5065583 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200001468.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Atp6v1c2

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transport
ATP hydrolysis coupled proton transport
proton transport
positive regulation of Wnt signaling pathway
hydrogen ion transmembrane transport
hydrogen-exporting ATPase activity, phosphorylative mechanism
hydrogen ion transmembrane transporter activity
protein dimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200010324.mm.1 NM_001159632.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200010324.mm.1 NM_001159632.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200010324.mm.1 NM_133699.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200010324.mm.1 NM_133699.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200010324.mm.1 TC1200001468.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0