This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00504670_m1
See other KHDRBS2 GE Assays ›
Gene Symbol:
Khdrbs2
Gene Name:
KH domain containing, RNA binding, signal transduction associated 2
Gene Aliases:
6330586C16Rik, Slim1, Slm1
Chromosome Location:
Chr.1: 32172688 - 32705224 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00504670_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Khdrbs2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133235.2 NP_573498.1 4 - 5 594 77
GenBank mRNA AK078160.1 - 4 - 5 673 77
BC132117.1 - 4 - 5 584 40130394 77
BC132119.1 - 4 - 5 584 40130396 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100000201.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Khdrbs2

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
positive regulation of signal transduction
nucleic acid binding
RNA binding
SH3/SH2 adaptor activity
poly(A) binding
poly(U) RNA binding
SH3 domain binding
SH2 domain binding
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100001197.mm.1 NM_133235.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100001197.mm.1 TC0100000201.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0