This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00504642_m1
See other PFKFB3 GE Assays ›
Gene Symbol:
Pfkfb3
Gene Name:
6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3
Gene Aliases:
E330010H22Rik, iPFK-2, uPFK-2
Chromosome Location:
Chr.2: 11471430 - 11554071 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00504642_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pfkfb3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001177752.1 NP_001171223.1 2 - 3 595 68
NM_001177753.1 NP_001171224.1 2 - 3 595 68
NM_001177754.1 NP_001171225.1 2 - 3 595 68
NM_001177755.1 NP_001171226.1 2 - 3 595 68
NM_001177756.1 NP_001171227.1 2 - 3 595 68
NM_001177757.1 NP_001171228.1 2 - 3 595 68
NM_001177758.1 NP_001171229.1 2 - 3 232 68
NM_133232.3 NP_573495.2 2 - 3 595 68
GenBank mRNA AF294617.1 - 2 - 3 530 68
AK054285.1 - 2 - 3 580 68
AK077210.1 - 2 - 3 181 68
AK138807.1 - 2 - 3 380 68
AK143330.1 - 2 - 3 202 68
AK154607.1 - 2 - 3 545 68
BC052400.1 - 2 - 3 572 5721049 68
BC057320.1 - 2 - 3 547 6826100 68
EU034675.1 - 2 - 3 205 68
EU034676.1 - 2 - 3 205 68
EU034677.1 - 2 - 3 205 68
EU034678.1 - 2 - 3 205 68
EU034679.1 - 2 - 3 205 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002961.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pfkfb3

Panther Classification:

Molecular Function -

carbohydrate phosphatase

Gene Ontology Categories:

Function(s) Process(es)

fructose 2,6-bisphosphate metabolic process
6-phosphofructo-2-kinase activity
fructose-2,6-bisphosphate 2-phosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0