This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00504219_m1
See other KANK3 GE Assays ›
Gene Symbol:
Kank3
Gene Name:
KN motif and ankyrin repeat domains 3
Gene Aliases:
0610013D04Rik, Ankrd47, D17Ertd288e, NG28
Chromosome Location:
Chr.17: 33810523 - 33822920 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00504219_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Kank3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030697.2 NP_109622.1 1 - 2 40 87
GenBank mRNA AK002623.1 - 1 - 2 41 87
AK132011.1 - 1 - 2 41 87
BC061055.1 - 1 - 2 40 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000593.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK002623.1,AK132011.1,BC061055.1,NM_030697.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Kank3

Panther Classification:

Molecular Function -

non-motor actin binding protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of stress fiber assembly
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700005428.mm.1 NM_030697.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700005428.mm.1 NM_030697.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700005428.mm.1 TC1700000593.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700005426.mm.1 NM_030697.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700005426.mm.1 NM_030697.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700005426.mm.1 TC1700000593.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0