This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00503957_m1
See other NDUFS7 GE Assays ›
Gene Symbol:
Ndufs7
Gene Name:
NADH dehydrogenase (ubiquinone) Fe-S protein 7
Gene Aliases:
1010001M04Rik
Chromosome Location:
Chr.10: 80249121 - 80256793 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00503957_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufs7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029272.3 NP_083548.1 1 - 2 42 63
GenBank mRNA AK003132.1 - 1 - 2 43 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000847.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK003132.1,NM_029272.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndufs7

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

transport
mitochondrial respiratory chain complex I assembly
oxidation-reduction process
protease binding
NADH dehydrogenase activity
NADH dehydrogenase (ubiquinone) activity
oxidoreductase activity
metal ion binding
quinone binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000006409.mm.1 NM_029272.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000006409.mm.1 NM_029272.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000006409.mm.1 TC1000000847.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0