This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00503619_m1
See other CSNK1D GE Assays ›
Gene Symbol:
Csnk1d
Gene Name:
casein kinase 1, delta
Gene Aliases:
1200006A05Rik, AA409348, D930010H05Rik
Chromosome Location:
Chr.11: 120958010 - 120991333 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00503619_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Csnk1d
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027874.2 NP_082150.1 3 - 4 653 121
NM_139059.2 NP_620690.1 3 - 4 653 121
GenBank mRNA AK004606.1 - 3 - 4 413 121
AK051052.1 - 3 - 4 652 121
AK088642.1 - 3 - 4 388 121
AK140317.1 - 3 - 4 378 121
AK143534.1 - 3 - 4 353 121
AK146600.1 - 3 - 4 643 121
AK150387.1 - 3 - 4 381 121
AK150815.1 - 3 - 4 380 121
AK152721.1 - 3 - 4 595 121
AK157812.1 - 3 - 4 388 121
AK199003.1 - 1 - 2 112 122
BC004604.1 - 3 - 4 639 3585286 121

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100004236.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Csnk1d

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
protein phosphorylation
endocytosis
regulation of cell shape
Wnt signaling pathway
phosphorylation
peptidyl-serine phosphorylation
positive regulation of proteasomal ubiquitin-dependent protein catabolic process
circadian regulation of gene expression
regulation of circadian rhythm
rhythmic process
spindle assembly
positive regulation of canonical Wnt signaling pathway
nucleotide binding
glycoprotein binding
protein kinase activity
protein serine/threonine kinase activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
peptide binding
tau-protein kinase activity
phosphoprotein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100040634.mm.1 NM_027874.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100040634.mm.1 NM_027874.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100040634.mm.1 NM_139059.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100040634.mm.1 NM_139059.2 Refseq
Affymetrix Mouse Clariom S Array PSR1100040634.mm.1 TC1100004236.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0