This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00503273_m1
See other TMBIM4 GE Assays ›
Gene Symbol:
Tmbim4
Gene Name:
transmembrane BAX inhibitor motif containing 4
Gene Aliases:
0610007H07Rik, AU022431
Chromosome Location:
Chr.10: 120208826 - 120224897 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00503273_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tmbim4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026617.3 NP_080893.1 4 - 5 365 106
GenBank mRNA AK006207.1 - 4 - 5 366 106
AK208947.1 - 2 - 3 96 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000001479.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tmbim4

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
negative regulation of apoptotic process
regulation of calcium-mediated signaling
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000010506.mm.1 NM_026617.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000010506.mm.1 NM_026617.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000010506.mm.1 TC1000001479.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0