This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00503056_m1
See other MFSD1 GE Assays ›
Gene Symbol:
Mfsd1
Gene Name:
major facilitator superfamily domain containing 1
Gene Aliases:
1200003O06Rik
Chromosome Location:
Chr.3: 67582768 - 67604231 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00503056_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mfsd1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025813.3 NP_080089.1 7 - 8 698 67
GenBank mRNA AK004586.1 - 7 - 8 699 67
AK032688.1 - 7 - 8 666 67
AK075859.1 - 7 - 8 728 67
AK135235.1 - 7 - 8 699 67
AK136108.1 - 7 - 8 690 67
AK152444.1 - 7 - 8 720 67
AK159554.1 - 7 - 8 726 67
BC024891.1 - 7 - 8 680 3992247 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300000555.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mfsd1

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
transmembrane transport
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300003764.mm.1 NM_025813.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300003764.mm.1 NM_025813.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300003764.mm.1 TC0300000555.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300003763.mm.1 NM_025813.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300003763.mm.1 NM_025813.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300003763.mm.1 TC0300000555.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0