This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00502790_m1
See other SLC38A10 GE Assays ›
Gene Symbol:
Slc38a10
Gene Name:
solute carrier family 38, member 10
Gene Aliases:
1810073N04Rik
Chromosome Location:
Chr.11: 120103951 - 120151351 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00502790_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc38a10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001164798.1 NP_001158270.1 6 - 7 903 64
NM_001164799.1 NP_001158271.1 6 - 7 903 64
NM_001164800.1 NP_001158272.1 6 - 7 903 64
NM_001164801.1 NP_001158273.1 6 - 7 903 64
NM_001164802.1 NP_001158274.1 6 - 7 903 64
NM_024249.5 NP_077211.4 6 - 7 903 64
GenBank mRNA AK007977.1 - 6 - 7 793 64
AK032245.1 - 6 - 7 827 64
AK090050.1 - 6 - 7 879 64
AK159948.1 - 6 - 7 865 64
AK165672.1 - 6 - 7 868 64
BC082300.1 - 6 - 7 874 30362204 64
BC088811.1 - 6 - 7 839 6493546 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100004196.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc38a10

Panther Classification:

Molecular Function -

amino acid transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
transport
ion transport
sodium ion transport
amino acid transport
bone development
molecular_function
amino acid transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100039952.mm.1 NM_001164798.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100039952.mm.1 NM_001164798.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100039952.mm.1 NM_001164799.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100039952.mm.1 NM_001164799.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100039952.mm.1 NM_001164800.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100039952.mm.1 NM_001164800.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100039952.mm.1 NM_001164801.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100039952.mm.1 NM_001164801.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100039952.mm.1 NM_001164802.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100039952.mm.1 NM_001164802.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100039952.mm.1 NM_024249.5.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100039952.mm.1 NM_024249.5 Refseq
Affymetrix Mouse Clariom S Array PSR1100039952.mm.1 TC1100004196.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0