This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00501522_m1
See other YES1 GE Assays ›
Gene Symbol:
Yes1
Gene Name:
Yamaguchi sarcoma viral (v-yes) oncogene homolog 1
Gene Aliases:
Yes, p61-Yes
Chromosome Location:
Chr.5: 32611171 - 32687066 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00501522_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Yes1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_009535.3 NP_033561.1 1 - 2 663 95
GenBank mRNA AK035612.1 - 1 - 2 650 95
AK167420.1 - 1 - 2 338 95
X67677.1 - 1 - 2 590 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500000344.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK035612.1,AK167420.1,NM_009535.3,X67677.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Yes1

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
glucose transport
phosphorylation
peptidyl-tyrosine phosphorylation
cell differentiation
cellular response to platelet-derived growth factor stimulus
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
innate immune response
positive regulation of transcription from RNA polymerase II promoter
protein autophosphorylation
positive regulation of peptidyl-tyrosine phosphorylation
cellular response to retinoic acid
cellular response to peptide hormone stimulus
cellular response to transforming growth factor beta stimulus
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
receptor binding
epidermal growth factor receptor binding
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
enzyme binding
ion channel binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500003140.mm.1 NM_009535.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003140.mm.1 NM_009535.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500003140.mm.1 TC0500000344.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500003142.mm.1 NM_001205132.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003142.mm.1 NM_001205132.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003142.mm.1 NM_001205133.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003142.mm.1 NM_001205133.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003142.mm.1 NM_009535.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003142.mm.1 NM_009535.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500003142.mm.1 TC0500000344.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500003137.mm.1 NM_001205132.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003137.mm.1 NM_001205132.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003137.mm.1 NM_001205133.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003137.mm.1 NM_001205133.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003137.mm.1 NM_009535.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003137.mm.1 NM_009535.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500003137.mm.1 TC0500000344.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500003138.mm.1 NM_001205132.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003138.mm.1 NM_001205132.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003138.mm.1 NM_001205133.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003138.mm.1 NM_001205133.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003138.mm.1 NM_009535.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500003138.mm.1 NM_009535.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500003138.mm.1 TC0500000344.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 873863 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0