This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00501013_g1
See other PDE7A GE Assays ›
Gene Symbol:
Pde7a
Gene Name:
phosphodiesterase 7A
Gene Aliases:
AU015378, AW047537
Chromosome Location:
Chr.3: 19223108 - 19311374 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00501013_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde7a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001122759.2 NP_001116231.1 8 - 9 1151 98
NM_008802.3 NP_032828.2 8 - 9 881 98
GenBank mRNA AK036517.1 - 8 - 9 839 98
AK154521.1 - 8 - 9 968 98
AK168957.1 - 8 - 9 843 98
AY007702.1 - 8 - 9 830 98
BC062909.1 - 8 - 9 1150 6815827 98
U68171.1 - 8 - 9 829 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300001697.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pde7a

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

signal transduction
metabolic process
cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
phosphoric diester hydrolase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300013327.mm.1 NM_001122759.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300013327.mm.1 NM_008802.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300013327.mm.1 TC0300001697.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0