This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00499731_m1
See other SLC45A2 GE Assays ›
Gene Symbol:
Slc45a2
Gene Name:
solute carrier family 45, member 2
Gene Aliases:
Aim-1, Aim1, Dbr, Matp, Oca4, blanc-sale, bls, uw
Chromosome Location:
Chr.15: 10995693 - 11029233 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00499731_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc45a2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053077.3 NP_444307.1 5 - 6 1252 106
GenBank mRNA AF360357.1 - 5 - 6 1160 106
AK029155.1 - 5 - 6 1253 106
AK089932.1 - 5 - 6 1223 106
AK148207.1 - 1250 106
AY034377.1 - 5 - 6 1173 106
BC125376.1 - 5 - 6 1239 40129891 106
BC132429.1 - 5 - 6 1239 40130706 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500000088.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Slc45a2

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s)

visual perception
developmental pigmentation
response to stimulus

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500000846.mm.1 NM_053077.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500000846.mm.1 NM_053077.3 Refseq
Affymetrix Mouse Clariom S Array PSR1500000846.mm.1 TC1500000088.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0