This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00499676_m1
See other IDH3A GE Assays ›
Gene Symbol:
Idh3a
Gene Name:
isocitrate dehydrogenase 3 (NAD+) alpha
Gene Aliases:
1110003P10Rik, 1500012E04Rik, AA407078, AI316514
Chromosome Location:
Chr.9: 54586511 - 54604662 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00499676_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Idh3a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029573.2 NP_083849.1 5 - 6 488 74
GenBank mRNA AK003393.1 - 2 - 3 381 74
AK010065.1 - 5 - 6 512 74
AK032787.1 - 5 - 6 512 74
AK047951.1 - 5 - 6 510 74
AK117103.1 - 5 - 6 524 74
AK146717.1 - 2 - 3 352 74
AK150618.1 - 5 - 6 519 74
AK151304.1 - 5 - 6 513 74
AK152353.1 - 5 - 6 522 74
AK153459.1 - 5 - 6 513 74
AK158646.1 - 5 - 6 666 74
AK159051.1 - 5 - 6 585 74
AK159666.1 - 5 - 6 519 74
AK160054.1 - 5 - 6 519 74
AK164038.1 - 6 - 7 614 74
AK167822.1 - 5 - 6 518 74
AK168034.1 - 5 - 6 510 74
AK168049.1 - 5 - 6 513 74
AK168149.1 - 5 - 6 519 74
AK169152.1 - 5 - 6 486 74
BC034273.1 - 5 - 6 478 4166193 74
BC049956.1 - 5 - 6 488 4212315 74

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Idh3a

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

tricarboxylic acid cycle
isocitrate metabolic process
2-oxoglutarate metabolic process
NADH metabolic process
oxidation-reduction process
magnesium ion binding
isocitrate dehydrogenase (NAD+) activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
metal ion binding
NAD binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0