This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00499599_m1
See other PRPF31 GE Assays ›
Gene Symbol:
Prpf31
Gene Name:
PRP31 pre-mRNA processing factor 31 homolog (yeast)
Gene Aliases:
1500019O16Rik, 2810404O06Rik, AW554706, PRP31, RP11
Chromosome Location:
Chr.7: 3629985 - 3642485 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00499599_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prpf31
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027328.4 NP_081604.3 9 - 10 1031 68
GenBank mRNA AK005294.1 - 9 - 10 999 68
AK033190.1 - 9 - 10 1034 68
AK033283.1 - 9 - 10 996 68
AK051260.1 - 9 - 10 991 68
AY040823.1 - 9 - 10 1007 68
BC018376.1 - 9 - 10 997 3673893 68
BC057877.1 - 8 - 9 974 5347609 68
CT010189.1 - 8 - 9 951 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000015.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prpf31

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

spliceosomal tri-snRNP complex assembly
mRNA splicing, via spliceosome
mRNA processing
RNA splicing
RNA binding
ribonucleoprotein complex binding
poly(A) RNA binding
snRNP binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700000153.mm.1 NM_001159714.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700000153.mm.1 NM_001159714.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700000153.mm.1 NM_027328.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700000153.mm.1 NM_027328.4 Refseq
Affymetrix Mouse Clariom S Array PSR0700000153.mm.1 TC0700000015.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0