This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00498691_g1
See other ITGAX GE Assays ›
Gene Symbol:
Itgax
Gene Name:
integrin alpha X
Gene Aliases:
AI449405, Cd11c, Cr4, N418
Chromosome Location:
Chr.7: 128129529 - 128150657 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00498691_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Itgax
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021334.2 NP_067309.1 7 - 8 760 69
GenBank mRNA AF211864.1 - 7 - 8 760 69
AK131133.1 - 7 - 8 766 69
AK155816.1 - 7 - 8 782 69
AK170432.1 - 7 - 8 778 69
BC042840.1 - 7 - 8 759 69
BC057200.1 - 7 - 8 753 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700001876.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Itgax

Panther Classification:

Molecular Function -

integrin

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
integrin-mediated signaling pathway
heterotypic cell-cell adhesion
activated T cell proliferation
defense response to virus
positive regulation of protein targeting to mitochondrion
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0