This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00498564_g1
See other NELFCD GE Assays ›
Gene Symbol:
Nelfcd
Gene Name:
negative elongation factor complex member C/D, Th1l
Gene Aliases:
2410003I03Rik, C77797, Th1, Th1l
Chromosome Location:
Chr.2: 174415804 - 174427503 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00498564_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nelfcd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020580.3 NP_065605.2 12 - 13 1538 77
GenBank mRNA AJ238741.1 - 12 - 13 1472 77
AJ238742.1 - 12 - 13 1538 77
AJ238743.1 - 12 - 13 1492 77
AK010360.1 - 12 - 13 1485 77
AK049418.1 - 12 - 13 1510 77
AK159866.1 - 12 - 13 1539 77
BC006959.1 - 12 - 13 1499 3707929 77
BC031747.1 - 12 - 13 1496 3673038 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200002705.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nelfcd

Gene Ontology Categories:

Function(s)

transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of transcription, DNA-templated

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200022149.mm.1 NM_020580.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200022149.mm.1 NM_020580.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200022149.mm.1 TC0200002705.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0