This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00497199_m1
See other ZEB2 GE Assays ›
Gene Symbol:
Zeb2
Gene Name:
zinc finger E-box binding homeobox 2
Gene Aliases:
9130203F04Rik, D130016B08Rik, SIP1, Zfhx1b, Zfx1b, Zfxh1b
Chromosome Location:
Chr.2: 44983512 - 45113279 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00497199_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Zeb2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289521.1 NP_001276450.1 8 - 9 3258 86
NM_015753.4 NP_056568.2 9 - 10 3595 86
GenBank mRNA AF033116.1 - 8 - 9 3110 86
AK032970.1 - 2 - 3 396 86
AK122312.1 - 9 - 10 3168 86
AK141157.1 - 8 - 9 3226 86
BC060699.1 - 9 - 10 3332 6408845 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200003473.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Zeb2

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
neural crest cell migration
somitogenesis
neural tube closure
transcription, DNA-templated
regulation of transcription, DNA-templated
central nervous system development
hippocampus development
cell proliferation in forebrain
positive regulation of Wnt signaling pathway
positive regulation of JUN kinase activity
positive regulation of melanocyte differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of melanin biosynthetic process
developmental pigmentation
embryonic morphogenesis
melanocyte migration
positive regulation of lens fiber cell differentiation
regulation of melanosome organization
nucleic acid binding
DNA binding
metal ion binding
R-SMAD binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200029711.mm.1 NM_001289521.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200029711.mm.1 NM_015753.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200029711.mm.1 TC0200003473.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200029716.mm.1 NM_001289521.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200029716.mm.1 NM_015753.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200029716.mm.1 TC0200003473.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200029714.mm.1 NM_001289521.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200029714.mm.1 NM_015753.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200029714.mm.1 TC0200003473.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200029713.mm.1 NM_001289521.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200029713.mm.1 NM_015753.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200029713.mm.1 TC0200003473.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200029715.mm.1 NM_001289521.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200029715.mm.1 NM_015753.4 Refseq
Affymetrix Mouse Clariom S Array PSR0200029715.mm.1 TC0200003473.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0